Learn More
Invitrogen™ Chargeswitch™ Pro Filter Plasmid Mini Kit
$171.00 - $489.00
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
CS31102 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
CS31102
N/A 10 preps
Each for $171.00
Add to cart
CS31103 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
CS31103
N/A 100 preps
Each for $489.00
Add to cart
Description
Description
The Chargeswitch™ -Pro Filter Plasmid Mini Kit includes 10 plasmid minipreps and provides high yield and high purity plasmid with the fastest and the simplest protocol.
The ChargeSwitch™-Pro Plasmid Filter Mini, Midi, Maxi Kits generates high-quality plasmid DNA without the use of harsh or potentially interfering reagents, for better success in downstream applications. The system is based upon a novel dual⁄nested column design which facilitate the fastest and simplest protocol for high quality plasmid. The system is composed of two columns. The inner flat bottomed column, also referred to as “lysate clarification” column, contains filter materials suitable for rapid clarification of bacterial lysate. The outer luer bottomed column, also referred to as “binding” column, contains ChargeSwitch™ (CST) derivatized membrane that immediately binds plasmid DNA from the clarified lysate. This nested-column design enables simultaneous clarification and DNA binding in a single short centrifugation (or vacuum) step. The combined advantages of novel, optimized plastics and a unique chemistry are an extreme competitive advantage of speed and convenience.
To accomplish this, the ChargeSwitch™-Pro Filter Kit provides:
  • 100% ethanol-free, guanidinium-free, organics-free, aqueous protocol for a safer, more streamlined method
  • Cleaner purified product for improved performance in downstream applications
  • A novel Dual⁄nested column design for simultaneous lysate clearing and binding step which offers the fastest and the simplest protocol
  • Endotoxin units < 10 EU ⁄ ug DNA for
  • Vacuum Elution can be done with the EveryPrep™ Universal Vacuum Manifold (Cat. No. K211101)
    How it works
    The ChargeSwitch™ technology replaces the use of ethanol, chaotropic salts, or organic solvents for nucleic acid purification with a simple ion-exchange mechanism. At low pH values (<6.5) the ChargeSwitch™ surface ligand is positively charged and binds plasmid DNA. Raising the pH (>8.5) neutralizes the charge and elutes the bound nucleic acid. The ChargeSwitch™ protocol retains the ease and familiarity of the common silica spin column protocol with a binding, washing, and elution step, making adapting to the ChargeSwitch™ method effortless.
  • Safety and Handling