Learn More
GSC Go Science Crazy Charles' Law Demonstration
Study the degree of expansion as it relates to temperature with this high quality Charle's Law.
Manufacturer:  GSC Go Science Crazy CHRLW01
Catalog No. S24659
Add to cart
Description
Description
The Charles' Law Apparatus is used to study the degree of expansion that is directly proportional to the increase in temperature. This Charles' Law Apparatus enables students to precisely determine the volume coefficient of expansion of air throughout a temperature range. By adding or draining the solution (though a stopclip located at the bottom of the tube), the solution in the tubes can be maintained at even levels while the temperature in the water jacket is varied. Temperature is changed by adding hot water though a funnel at the top of the jacket, as cooler water is drained from below though a rubber tube.