Learn More
CHCHD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 210901AP
Catalog No. 50-173-3252
Add to cart
Description
Description
CHCHD4, a component of human mitochondria, belongs to a protein family whose members share 6 highly conserved cysteine residues constituting a -CXC-CX(9)C-CX(9)C- motif in the C terminus.
Specifications
Specifications
CHCHD4
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N4Q1, Q8VEA4
CHCHD4, MIA40
Rabbit
IgG
150 μL
-20°C
Primary
131474, 312559, 72170
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
CHCHD4
Liquid
Chchd4
CHCHD4 Fusion Protein Ag15339
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat