Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHCHD5 Mouse anti-Human, Clone: OTI3H3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525411

Catalog No. PIMA525411


Add to cart

Description

Description

CHCHD5 (Coiled-Coil-Helix-Coiled-Coil-Helix Domain Containing 5) is a Protein Coding gene. Among its related pathways are Metabolism of proteins and Mitochondrial protein import.
Specifications

Specifications

CHCHD5
Monoclonal
0.35 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BSY4
C2orf9
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84269
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI3H3
Unconjugated
CHCHD5
Liquid
CHCHD5
Human recombit protein fragment corresponding to amino acids 1-110 of CHCHD5 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit