Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHCHD6 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP179820

 View more versions of this product

Catalog No. NBP179820


Add to Cart

Description

Description

CHCHD6 Polyclonal specifically detects CHCHD6 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CHCHD6
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
coiled-coil-helix-coiled-coil-helix domain containing 6, coiled-coil-helix-coiled-coil-helix domain-containing protein 6, MGC13016
Rabbit
26 kDa
100 μL
Primary
Expected identity based on immunogen sequence: Pig: 77%; Mouse: 77%; Guinea pig: 77%.
Human, Mouse, Guinea Pig
IgG
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
NP_115719
CHCHD6
Synthetic peptide directed towards the middle region of human CHCHD6The immunogen for this antibody is CHCHD6. Peptide sequence AAIQDKLFQVAKREREAATKHSKASLPTGEGSISHEEQKSVRLARELESR.
Affinity purified
RUO
84303
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit