Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

CHCHD6 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$152.22 - $349.40

Specifications

Antigen CHCHD6
Immunogen Synthetic peptide directed towards the middle region of human CHCHD6The immunogen for this antibody is CHCHD6. Peptide sequence AAIQDKLFQVAKREREAATKHSKASLPTGEGSISHEEQKSVRLARELESR.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP17982020
View Documents
Novus Biologicals
NBP17982020UL
20ul
Each for $152.22
N/A
NBP179820
View Documents
Novus Biologicals
NBP179820
100 ul
Each for $349.40
N/A
Description

Description

CHCHD6 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

CHCHD6
Rabbit
Polyclonal
Unconjugated
84303
Affinity Purified
Western Blot
RUO
Synthetic peptide directed towards the middle region of human CHCHD6The immunogen for this antibody is CHCHD6. Peptide sequence AAIQDKLFQVAKREREAATKHSKASLPTGEGSISHEEQKSVRLARELESR.
Primary
IgG
coiled-coil-helix-coiled-coil-helix domain containing 6, coiled-coil-helix-coiled-coil-helix domain-containing protein 6, MGC13016
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit