Learn More
CHD2 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer: Abnova Corporation H00001106T01
Description
Western Blotting
Specifications
293T | |
NM_001042572.1 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-CHD2 antibody (H00001106-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Human |
CHD2 293T Cell Transient Overexpression Lysate(Denatured) | |
56.8 | |
pCMV-CHD2 full-length | |
100 uL | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Western Blot |