Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen CHD2
Applications Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB096169
View Documents
Novus Biologicals
NBP304885100UL
100 μg Each for $320.71
Add to cart
 
NB096170
View Documents
Novus Biologicals
NBP30488520UL
20 μg Each for $138.37
Add to cart
 
Description

Description

CHD2 Polyclonal antibody specifically detects CHD2 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

CHD2
Unconjugated
Rabbit
Chromatin Research
PBS with 50% glycerol, pH7.3.
1106
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence
Purified
RUO
Human
ATP-dependent helicase CHD2, CHD-2, chromodomain helicase DNA binding protein 2, chromodomain-helicase-DNA-binding protein 2, DKFZp547I1315, DKFZp686E01200, DKFZp781D1727, EC 3.6.1, EC 3.6.4.12, FLJ38614
Recombinant fusion protein containing a sequence corresponding to amino acids 242-501 of human CHD2 (NP_001036037.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit