Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHD3 Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™
Mouse Recombinant Monoclonal Antibody
$100.30 - $454.30
Specifications
Antigen | CHD3 |
---|---|
Concentration | 1 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3794
|
Active Motif
91274 |
10 μg |
Each for $100.30
|
|
|||||
50-199-3793
|
Active Motif
91273 |
100 μg |
Each for $454.30
|
|
|||||
Description
This gene encodes a member of the CHD family of proteins which are characterized by the presence of chromo domains and SNF2-related helicase/ATPase domains. This protein is one of the components of a histone deacetylase complex referred to as the Mi-2/NuRD complex which participates in the remodeling of chromatin by deacetylating histones. Chromatin remodeling is essential for many processes including transcription. Autoantibodies against this protein are found in a subset of patients with dermatomyositis. Three alternatively spliced transcripts encoding different isoforms have been described.Specifications
CHD3 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Drosophila | |
O16102 | |
40111 | |
This antibody was raised against a synthetic peptide corresponding to amino acid residues 865-885 of Drosophila. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
CHD3 |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5 | |
2600010P09Rik; AF020312; ATP-dependent helicase CHD3; CHD3; CHD-3; Chd7; chromodomain helicase DNA binding protein 3; Chromodomain-helicase-DNA-binding protein 3; hZFH; MGC40857; Mi 2a; Mi2 ALPHA; Mi-2 alpha; Mi-2 autoantigen 240 kDa protein; Mi-2a; Mi2-alpha; Prp7; Prp9 1; Prp9-1; ZFH; Zinc finger helicase; zinc-finger helicase (Snf2-like) | |
CHD3 | |
IgG2a | |
Protein A | |
Antibody |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title