Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD3 Mouse anti-Human, Mouse, Clone:, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524760

Catalog No. PIMA524760

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a member of the CHD family of proteins which are characterized by the presence of chromo domains and SNF2-related helicase/ATPase domains. This protein is one of the components of a histone deacetylase complex referred to as the Mi-2/NuRD complex which participates in the remodeling of chromatin by deacetylating histones. Chromatin remodeling is essential for many processes including transcription. Autoantibodies against this protein are found in a subset of patients with dermatomyositis. Three alternatively spliced transcripts encoding different isoforms have been described.
Specifications

Specifications

CHD3
Monoclonal
ascites with 0.02% sodium azide
Q12873
226;2600010P09Rik;AF020312;ATP dependent helicase CHD3;ATP-dependent helicase Chd3;CHD-3;Chd3;CHD3_HUMAN;Chd7;Chromodomain-helicase-DNA-binding protein 3;hZFH;MGC40857;Mi 2 autoantigen 240 kDa protein;Mi 2a;Mi-2 autoantigen 240 kDa protein;Mi2 ALPHA;Mi2-alpha; Prp7;Prp9 1;ZFH;Zinc finger helicase;zinc-finger helicase (Snf2-like).
Mouse
IgG1
RUO
Antibody
Monoclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
CHD3
Liquid
CHD3
human CHD3 recombit protein
100 μL
-20°C
Primary
1107, 216848
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit