Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHD6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540944
Description
Peptide sequence: SCTDSAARTK SRKASKEQGP TPVEKKKKGK RKSETTVESL ELDQGLTNPS Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.
This gene encodes a member of the SNF2/RAD54 helicase protein family. The encoded protein contains two chromodomains, a helicase domain, and an ATPase domain. Several multi-subunit protein complexes remodel chromatin to allow patterns of cell type-specific gene expression, and the encoded protein is thought to be a core member of one or more of these chromatin remodeling complexes. The encoded protein may function as a transcriptional repressor and is involved in the cellular repression of influenza virus replication.
Specifications
CHD6 | |
Polyclonal | |
Unconjugated | |
CHD6 | |
5430439G14Rik; 6330406J24Rik; ATP-dependent helicase CHD6; CHD5; Chd6; CHD-6; chromodomain helicase DNA binding protein 6; chromodomain-helicase-DNA-binding protein 6; helicase C-terminal domain- and SNF2 N-terminal domain-containing protein; Kiaa1335; Radiation-induced gene B protein; RIGB | |
Rabbit | |
Affinity chromatography | |
RUO | |
84181 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TD26 | |
CHD6 | |
Synthetic peptide directed towards the C-terminal of human CHD6 (aa 171-220). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction