Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHD8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568145
Description
CHD8 Polyclonal Antibody for Western Blot
DNA helicase that acts as a chromatin remodeling factor and regulates transcription. Acts as a transcription repressor by remodeling chromatin structure and recruiting histone H1 to target genes. Suppresses p53/TP53-mediated apoptosis by recruiting histone H1 and preventing p53/TP53 transactivation activity. Acts as a negative regulator of Wnt signaling pathway by regulating beta-catenin (CTNNB1) activity. Negatively regulates CTNNB1-targeted gene expression by being recruited specifically to the promoter regions of several CTNNB1 responsive genes. Involved in both enhancer blocking and epigenetic remodeling at chromatin boundary via its interaction with CTCF. Acts as a suppressor of STAT3 activity by suppressing the LIF-induced STAT3 transcriptional activity. Also acts as a transcription activator via its interaction with ZNF143 by participating in efficient U6 RNA polymerase III transcription.
Specifications
CHD8 | |
Polyclonal | |
Unconjugated | |
Chd8 | |
5830451P18Rik; ATP-dependent helicase CHD8; AU015341; AUTS18; Axis duplication inhibitor; beta-catenin binding protein; CHD8; CHD-8; chromodomain helicase DNA binding protein 8; chromodomain-helicase-DNA-binding protein 8; Duplin; helicase with SNF2 domain 1; HELSNF1; Kiaa1564; mKIAA1564 | |
Rabbit | |
affinity chromatography | |
RUO | |
57680 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9HCK8 | |
Chd8 | |
Synthesized peptide derived from human CHD8. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction