Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine-Free/Total

Accurate test kits for routine water analysis

Supplier:  Lovibond™ 147010

 View more versions of this product

Catalog No. 09-053-104


Only null left
Add to Cart

Includes:
  • Aqua Comparator Housing (D55 for L147000)
  • Aqua Comparator Disc
  • DPD No. 1 Rapid Tablets
  • DPD No. 3 Rapid Tablets
  • 2 x 13.5mm Sample Cells
  • Tablet Crusher
  • Instructions
  • Carrying case

Description

Description

Accurate test kits for routine water analysis

  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value

Specifications

Specifications

±5%
Aqua comparator™ housing, aqua comparator™ disc, DPD No. 1 rapid tablets, DPD No. 3 rapid tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
0 to 1mg/L
Routine water analysis
100
Chlorine Free and Total Test Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.