Learn More
OHAUS CORPORATION CHECKWEIGH SCALE
Combining an NSF-certified and USDA accepted food-safe base, durable stainless steel construction, and industry leading software, the OHAUS CKW Series includes fully-dedicated checkweighing scales built for applications requiring fast and accurate operation.
Manufacturer:  OHAUS CORPORATION 30448474
 View more versions of this product
Catalog No. 02-115-02
Add to cart
Description
Description
User-configurable resolution up to 7,500d/3,000e, 128 part library, 3-color checkweighing LEDS, beeper, alibi memory kit (sold separately as an accessory), 150% overload capacity protection, five operating languages, NTEP-certified and Measurement Canada approved
Specifications
Specifications
External calibration
LCD
AC Adapter
2 g
11 kg
15 kg
Yes
485 mm
390 mm
2 Seconds
410 mm
Bench Scale