Learn More
OHAUS™ CHECKWEIGH SCALE
Combining an NSF-certified and USDA accepted food-safe base, durable stainless steel construction, and industry leading software, the OHAUS CKW Series includes fully-dedicated checkweighing scales built for applications requiring fast and accurate operation.
Manufacturer:  OHAUS™ 30448472
 View more versions of this product
Catalog No. 02-115-00
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
User-configurable resolution up to 7,500d/3,000e, 128 part library, 3-color checkweighing LEDS, beeper, alibi memory kit (sold separately as an accessory), 150% overload capacity protection, five operating languages, NTEP-certified and Measurement Canada approved
Specifications
Specifications
External calibration
3 kg
0.5 g
2 sec.
390 mm
485 mm
Yes
LCD
AC Adapter
Bench Scale
410 mm
9 kg