Learn More
CHEK2 Mouse anti-Human, Clone: 73C175.1.1, Abnova™
Mouse monoclonal antibody raised against human CHEK2.
Manufacturer:  Abnova Corporation MAB12514
Catalog No. 89-933-968
Add to cart
Description
Description
In response to DNA damage and replication blocks, cell cycle progression is halted through the control of critical cell cycle regulators. The protein encoded by this gene is a cell cycle checkpoint regulator and putative tumor suppressor. It contains a forkhead-associated protein interaction domain essential for activation in response to DNA damage and is rapidly phosphorylated in response to replication blocks and DNA damage. When activated, the encoded protein is known to inhibit CDC25C phosphatase, preventing entry into mitosis, and has been shown to stabilize the tumor suppressor protein p53, leading to cell cycle arrest in G1. In addition, this protein interacts with and phosphorylates BRCA1, allowing BRCA1 to restore survival after DNA damage. Mutations in this gene have been linked with Li-Fraumeni syndrome, a highly penetrant familial cancer phenotype usually associated with inherited mutations in TP53. Also, mutations in this gene are thought to confer a predisposition to sarcomas, breast cancer, and brain tumors. This nuclear protein is a member of the CDS1 subfamily of serine/threonine protein kinases. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
Specifications
CHEK2
73C175.1.1
Mouse monoclonal antibody raised against human CHEK2.
In PBS (0.02% sodium azide).
CDS1/CHK2/HuCds1/LFS2/PP1425/RAD53
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
CHEK2
A synthetic peptide corresponding to human CHEK2.
Protein G purification
RUO
Primary
11200