Learn More
Crystal Chem IncSupplier Diversity Partner Chemerin ELISA Kit, 96-wells DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80589
The Chemerin ELISA is used to quantify levels of human chemerin in cell culture, plasma, serum , cerebrospinal fluid, urine, breast milk and amniotic fluid using only a 5 µL sample volume. Highlights: Kit uses sample size of only 10 µL. Dynamic range of 25 - 600 pg/mL. High sensitivity of 5 pg/mL. Designed for plasma, serum, cerebrospinal fluid, urine, breast milk, amniotic fluid or cell culture. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7755