Learn More
Chemerin Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0185-6G32, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212190AF532
Catalog No. NP212190A32
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Chemerin Monoclonal antibody specifically detects Chemerin in Human samples. It is validated for Western Blot.
Specifications
Specifications
Chemerin
MM0185-6G32
Western Blot
chemerin, RAR-responsive protein TIG2, retinoic acid receptor responder (tazarotene induced) 2, retinoic acid receptor responder protein 2, Tazarotene-induced gene 2 protein, TIG2HP10433
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
RARRES2
Human Chemerin
0.1 mL
Primary
5919