Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemerin Rat anti-Mouse, Alexa Fluor™ 532, Clone: 13G2, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP221989AF532

Catalog No. NP221989A32


Add to cart

Description

Description

Chemerin Monoclonal antibody specifically detects Chemerin in Mouse samples. It is validated for ELISA.
Specifications

Specifications

Chemerin
13G2
ELISA
chemerin, RAR-responsive protein TIG2, retinoic acid receptor responder (tazarotene induced) 2, retinoic acid receptor responder protein 2, Tazarotene-induced gene 2 protein, TIG2HP10433
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
ELISA
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
RARRES2
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein Chemerin. Rat monoclonal anti mouse Chemerin Q9DD06.
0.1 mL
Primary
5919
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit