Learn More
CHEMetrics™ Chemets™ Water Testing Kits
Provide results in two minutes or less
Manufacturer:  CHEMetrics™ K7501
 View more versions of this product
Catalog No. 13-299-201
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Dissolved Oxygen
0 to 1ppm
Plastic case, 30 ampoules of reagent, refill, comparator, 25mL sample cup, instructions, adhesive mounting clamp, permanent mounting clamp, sampling tube
<1 min.
Chemets™ Water Testing Kit
Water testing kits for chlorine (DPD), copper, dissolved oxygen, hydrogen peroxide, phenols, phosphate, silica, sulfide and more
30.0
Rhodazine D