CHEMetrics™ Chemets™ Water Testing Kits
Provide results in two minutes or less
Manufacturer: CHEMetrics™ K2505
 View more versions of this product
Catalog No. 13-299-215B
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Water testing kits for chlorine (DPD), copper, dissolved oxygen, hydrogen peroxide, phenols, phosphate, silica, sulfide and more
    No. of Tests 30.0
    Method DDPD
    Range 0 to 1, 1 to 5ppm
    Includes Plastic case, 30 ampoules of reagent, comparator, snap cup, instructions
    Analytes Free and Total Chlorine
    Product Type Chemets™ Water Testing Kit
  • Description & Specifications
Related Products
Product Content Feedback