CHEMetrics™ Chemets™ Water Testing Kits
Provide results in two minutes or less
Manufacturer: CHEMetrics™ K4520
 View more versions of this product
Catalog No. 13-299-216E
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Water testing kits for chlorine (DPD), copper, dissolved oxygen, hydrogen peroxide, phenols, phosphate, silica, sulfide and more
    Includes Cardboard box, 30 ampoules of reagent with valve assemblies, titrettor, 25mL sample cup, instructions
    No. of Tests 30.0
    Method Total EDTA
    Product Type Chemets™ Water Testing Kit
    Range 20 to 200ppm
  • Description & Specifications
Related Products
Product Content Feedback