Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Chemical Absorbent Pads
Color-coded absorbents for safe clean-up of hazardous fluids. Brady™ Chemical Absorbent Pads are surfactant treated used on wide range of chemicals.
Specifications
Certifications/Compliance | EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910.120(j)(1)(vii); OSHA 29 CFR 1910.22(a)(2) |
---|---|
Color | Yellow |
Material | Meltblown Polypropylene |
Type | Perforated Chemical Absorbent Pad |
Weight Capacity | Medium-weight |
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Price | Quantity & Availability | ||||
19-150-259
|
Brady™
CH1212 |
30 gal. | 113.5 L |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
19-103-103
|
Brady™
CH121250 |
15 gal. | 56.7 L |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
Description
- Made of MAXX technology
- Benefits of sign and an absorbent in one
- Highly absorbent for all-purpose industrial applications
- Ideal for areas where the risk of slips, trips and falls are greatest
- Chemically inert, do not react with aggressive fluids
- Perforations provide cost control and reduce waste
- Medium weight - optimal for most general maintenance needs
Specifications
EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910.120(j)(1)(vii); OSHA 29 CFR 1910.22(a)(2) | |
Meltblown Polypropylene | |
Medium-weight | |
12 in. | |
12 in. |
Yellow | |
Perforated Chemical Absorbent Pad | |
12 x 12 in. (30.48 x 30.48 cm) | |
30.48 cm | |
30.48 cm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title