Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Chemical Hazard Identification Labels
High-visibility red-and-white pressure-sensitive vinyl labels ideal for alerting employees to potential chemical hazards in the workplace
Supplier: National Marker™ HC 206
Description
- Withstand most chemicals, solvents and vapors
- Resist fading and scratching
- For medium-sized containers
- Measure 3 × 5 in.
Specifications
White, Red | |
Vinyl | |
Hazard identification, suitable for both indoor and outdoor applications | |
0.0045 mil | |
3 in. |
Label | |
Alcohol solution Container Label | |
10/Pk. | |
5 in. | |
7.62 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction