Learn More
Thermo Scientific™ Chemical Preservative Glass Ampoules
Save the time and tedium associated with diluting and aliquoting chemicals for environmental sample preservation by using pre-mixed Chemical Preservative Glass Ampoules.
$124.50
Specifications
Liner Material N/A
No. per Case 24
Certifications/Compliance Packaged in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Clas
Color Clear
Level None
View More Specs
Products 2
Catalog Number Mfr. No. Preservative Level No. per Case Price Quantity & Availability  
Catalog Number Mfr. No. Preservative Level No. per Case Price Quantity & Availability  
5005728285
View Documents
Thermo Scientific™
ACN51
5mL 1:1 Nitric Acid None 24 Case of 24 for $124.50
Add to cart
5005720396
View Documents
Thermo Scientific™
ACS2
2mL Sulfuric Acid, concentrated None 24 Case for $119.70
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Save the time and tedium associated with diluting and aliquoting chemicals for environmental sample preservation by using pre-mixed Thermo Scientific™ Chemical Preservative Glass Ampoules. Our pre-formulated and aliquoted glass ampoules are ready for field use. Just break the ampoule at the pre-scored neck area using the provided “breaker” tool, and pour into the designated sample bottle. Choose from our standard offering or let us custom-fill ampoules according to your personal analytical needs.
  • Glass ampoule containing a pre-measured aliquot of preservative chemical solution
  • Ampoules are machine-welded closed after filling for a consistent, leak-tight seal
  • Packaged and shipped in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Classification regulations
Ordering Information:
Custom preserved ampoules available by special order:
  • Choose a chemical from our standard chemical list
  • Specify the volume and concentration of chemical per ampoule
  • Certificate of chemical purity available upon request
  • 10 Case minimum order
Products available only in the US and Canada
Includes:
Preserved ampoule. MSDS available upon request.
Specifications
Specifications
N/A
Packaged in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Clas
None
24
Clear
Glass