Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chemical Reactions Lab Investigation
Explore how chemical reactions work and relate to both chemistry and biology.
Specifications
Class Size | 40 students |
---|---|
For Use With (Application) | To explore how chemical reactions work and relate to both chemistry and biology |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S64997
![]() |
Neo/Sci Corporation
201703 |
Chemical Reactions Lab Investigation Chemistry Kit |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Designed as a series of inquiry-based experiments
- Discover the definition of a chemical reaction
- Understand its nature, mechanism, properties and applications
- Demonstrate and observe the changes that accompany chemical reactions
- Witness changes in temperature, color, gas production and formation of precipitates Investigate a biochemical reaction
- Gain a better understanding of the importance and role of chemical reactions in the world around us Curriculum pack includes this kit and its corresponding, individual CD-ROM
- Class size: 40 students
Specifications
40 students | |
To explore how chemical reactions work and relate to both chemistry and biology |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title