Learn More
MilliporeSigma™ Chemicon™ Human Collagen Type II
Purified using differential salt precipitation, alcohol precipitation and DEAE chromatography
Supplier: MilliporeSigma™ CC052
Description
- COL2A1 is the gene responsible for the production of the alpha1(II) chain of type II collagen
Specifications
1 mg/mL | |
100 μg | |
Aliquot and freeze. Maintain at -20°C up to 12 months. Avoid repeated freeze/thaw cycles. |
Cell Culture | |
Collagen, Human Type II | |
Liquid |