Learn More
Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors Available on GSA/VA Contract for Federal Government customers only.
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves.
Manufacturer:  Thermo Scientific™ C21006
 View more versions of this product
Catalog No. 03-052-405
  Check Availability
Add to cart
Description
Description
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves. It can be actuated manually or with a microelectronic actuator. With 1/16 in. fittings and 0.010 in. ports, it is available in six-port or ten-port configurations and includes a 5µL sample loop.
  • 1/16 in. fittings
  • 0.010 in. ports
  • Available in 6-port or 10-port configurations
  • Available with manual or microelectric actuator
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
HPLC
Manual
6
5µL Loop
Manual Actuation
Model C2 Injector
N60 stainless
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.