Learn More
Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors Available on GSA/VA Contract for Federal Government customers only.
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves.
Manufacturer:  Thermo Scientific™ C21000EP
 View more versions of this product
Catalog No. 03-052-404
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves. It can be actuated manually or with a microelectronic actuator. With 1/16 in. fittings and 0.010 in. ports, it is available in six-port or ten-port configurations and includes a 5µL sample loop.
  • 1/16 in. fittings
  • 0.010 in. ports
  • Available in 6-port or 10-port configurations
  • Available with manual or microelectric actuator
Description & Specifications
Specifications
Microelectric
10
5µL Loop
Model C2 Injector
N60 stainless