Learn More
Thermo Scientific™ Cheminert™ Sample injector loops, PAEK, 5µL Available on GSA/VA Contract for Federal Government customers only.
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves.
Manufacturer:  Thermo Scientific™ CZSL5PK
Catalog No. 03-052-419
Add to cart
Description
Description
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves. It can be actuated manually or with a microelectronic actuator. With 1/16″ fittings and 0.010″ ports, it is available in six-port or ten-port configurations and includes a 5µL sample loop.
  • 1/16 in. fittings
  • 0.010 in. ports
  • Available in 6-port or 10-port configurations
  • Available with manual or microelectric actuator
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
5µL
Sample injector loops, PAEK