Learn More
Chemistry of Sugars Model Set Products for Science Education
Model provides a visual explanation of the chemistry of sugars.
Manufacturer:  Ryler Enterprises SUG1
Catalog No. S94027
Add to cart
Includes: Kit Contents:
  • 26 black 4-peg carbon atom centers
  • 26 red 2-peg oxygen atom centers
  • 50 white 1-peg hydrogen atom centers
  • 90-1.25 in. PVC, clear bonds (single bonds)
  • 10-1.625 in. PVC, clear bonds (double bonds)
  • Instructions for assembly
Description
Description
Students will gain an understanding of sugars through using this model to build structures and observe the bonds they form.
  • Color-coded, modeled plastic atom centers are 1/4 in. diameter
  • Pegs are 3/8 in. long and are set at angles peculiar to the elements the atoms represent
  • Standard color-coding
  • Atom centers are bonded by connecting pegs with pvc tubing
Specifications
Specifications
Understanding of sugars to build structures and observe the bonds they form
26 Black 4-peg Carbon atom centers, 26 red 2-peg Oxygen atom centers, 50 white 1-peg Hydrogen atom centers, Ninety 1.25 in. PVC clear bonds (single bonds), Ten 1.625 in. PVC clear bonds (double bonds), instructions for assembly
Chemistry of Sugar model set