Learn More
Lakeland Industries ChemMax 1 Serged Seam Coverall - Elastic Wrist/Ankle
Lakeland Industries Inc. ChemMax 1 Serged Seam Coverall - Elastic Wrist/Ankle offer entry level chemical protection for industry with excellent resistance to high and low pH chemicals
Manufacturer:  Lakeland Industries C1B417YLG
 View more versions of this product
Catalog No. 19-146-326
Add to cart
This item is not returnable. View return policy
Description
Description
  • ChemMax 1 chemical protection for general industry, environmental clean up and chemical handling
  • Serged seams for protection from light liquid splash.
Specifications
Specifications
Elastic
Elastic
Collared
Chemical
Large
Yellow
No
25/Cs.
Stitched, Bound