Learn More
Lakeland Industries ChemMax 1 Serged Seam Coverall - Elastic Wrist/Ankle
Lakeland Industries Inc. ChemMax 1 Serged Seam Coverall - Elastic Wrist/Ankle offer entry level chemical protection for industry with excellent resistance to high and low pH chemicals
Manufacturer: Lakeland Industries C1B417Y3X
This item is not returnable.
View return policy
Description
- ChemMax 1 chemical protection for general industry, environmental clean up and chemical handling
- Serged seams for protection from light liquid splash.
Specifications
Elastic | |
Elastic | |
Collared | |
Chemical | |
3X-Large |
Yellow | |
No | |
25/Cs. | |
Stitched, Bound |