Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ ChemPly™ N-740 Chemical Protection Polychloroprene Gloves

Gloves feature two superimposed neoprene layers to increase the mechanical and chemical resistance. Red inner layer ensures that any wear is clearly visible.

$314.00

Specifications

Certifications/Compliance 21 CFR 170-199
Chlorinated Yes
Color Black
Latex Free Yes
Material Neoprene
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-048-517L
View Documents
MAPA™
414709
9
Dozen of 12 for $314.00
Add to cart
 
19-048-517K
View Documents
MAPA™
414700
10
Dozen of 12 for $314.00
Add to cart
 
19-048-517J
View Documents
MAPA™
414701
11
Dozen of 12 for $314.00
Add to cart
 
Description

Description

  • Resistant to numerous solvents, oils, acids, alcohols and alkalis
  • Straight cuff design
  • Embossed pattern finish ensures secure grip
  • Chlorinated interior for easy donning and doffing
  • 18 in. length
  • 30 mil thickness
Specifications

Specifications

21 CFR 170-199
Black
Neoprene
0.76 mm
No
Straight
Unlined
18 in.
Yes
Yes
30 mil
EN 388:3112
Bacteria, Fungi only
Embossed
Chem-Ply™ N-740
45.7 cm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit