Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHES, JustPURE™, white powder, Fisher BioReagents

Click to view available options
Quantity:
100g
Packaging:
Poly Bottle
103-47-9
C8H17NO3S
207.288
MKWKNSIESPFAQN-UHFFFAOYSA-N
CHES, 2-(Cyclohexylamino)ethanesulfonic Acid
66898
2-(cyclohexylamino)ethanesulfonic acid
C1CCC(CC1)NCCS(=O)(=O)O
103-47-9
C8H17NO3S
207.288
MKWKNSIESPFAQN-UHFFFAOYSA-N
CHES, 2-(Cyclohexylamino)ethanesulfonic Acid
66898
2-(cyclohexylamino)ethanesulfonic acid
C1CCC(CC1)NCCS(=O)(=O)O
Purity 99% min.
Solubility Soluble in water
Absorbance 0.08 max. at 260nm
Aluminum (Al) 0.0005% max.
Ammonia 0.05% max.
Calcium (Ca) 0.001% max.
Chloride 0.05% max.
Assay Percent Range ≥99 %
Copper (Cu) 0.0005% max.
Formula Weight 207.29
Physical Form Solid
Grade JustPURE™
Lead (Pb) 0.001% max.
Insoluble Matter 0.1% max.
Iron (Fe) 0.0005% max.
Magnesium (Mg) 0.0005% max.
Packaging Poly Bottle
Phosphorus 0.0005% max.
Potassium (K) 0.005% max.
Residue after Ignition 0.1% max.
Sodium (Na) 0.01% max.
Sulfate 0.05% max.
Water 1% max.
Zinc (Zn) 0.0005% max.
Melting Point 300°C
Color White
Quantity 100g
Chemical Name or Material CHES
Show More Show Less

WARNING!

Emergency Overview
Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:2
Flammability:0
Instability:0