Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHGA Mouse anti-Human, Clone: CHGA/765, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14231

Catalog No. 89-034-315


Add to cart

Description

Description

Mouse monoclonal antibody raised against full length recombinant human CHGA.

  • High specificity
  • High reactivity
Specifications

Specifications

CHGA
CHGA/765
Mouse monoclonal antibody raised against full length recombinant human CHGA.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
CHGA
Recombinant protein corresponding to full length human CHGA.
Protein A/G purification
RUO
Primary
1113
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
P10645
CGA
Mouse
IgG2a, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)