Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHGA, Rabbit anti-Human, Clone: D25-R, Abnova™

Rabbit Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB15979

Catalog No. 89-998-686

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

CHGA
D25-R
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) The optimal working dilution should be determined by the end user.
chromogranin A (parathyroid secretory protein 1)
CGA
Rabbit
100 μL
Store at 4°C. Do not freeze.
Primary
1113
Immunohistochemistry (Paraffin)
Unconjugated
In 20 mM Tris-HCl buffer, pH 8.0 (20 mg/mL BSA, 0.05% Sodium Azide).
Liquid
CHGA
A synthetic peptide corresponding to N-terminus of human CHGA.
RUO
Antibody
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit