Learn More
CHGN 293T Cell Overexpression Lysate 2 (Denatured), Abnova
Manufacturer:  Abnova Corporation H00055790T02
Catalog No. 89-002-461
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
BC060772
6.8
Transient overexpression cell Lysate was tested with Anti-ChGn antibody (H00055790-B02) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
ChGn 293T Cell Transient Overexpression Lysate(Denatured)
32.78
pCMV-ChGn full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.