Learn More
Invitrogen™ CHI3L1/YKL-40 Human ELISA Kit
Sandwich ELISA Kit
$600.00 - $4840.00
Specifications
Accession Number | P36222 |
---|---|
Assay Sensitivity | 6 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EHCHI3L1
|
Invitrogen™
EHCHI3L1 |
96 Tests |
Pack for $600.00
|
|
|||||
EHCHI3L1X5
|
Invitrogen™
EHCHI3L1X5 |
5 x 96 Tests |
Each for $2,930.00
|
|
|||||
EHCHI3L1X10
|
Invitrogen™
EHCHI3L1X10 |
10 x 96 Tests |
Each for $4,840.00
|
|
|||||
Description
The Human CHI3L1/YKL-40 ELISA quantitates Hu CHI3L1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu CHI3L1. Principle of the method The Human CHI3L1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Human cartilage glycoprotein 39 (GP-39), also known as YKL-40, is a glycoprotein secreted by articular chondrocytes, synoviocytes and macrophages. Serum and synovial fluid GP-39 levels are elevated in inflammatory diseases and correlate with the degree of joint destruction in rheumatoid arthritis. GP-39 is expressed in articular chondrocytes and synovial cells, as well as in liver, but is undetectable in muscle tissues, lung, pancreas, mononuclear cells and fibroblasts. GP-39 is a candidate autoantigen in rheumatoid arthritis and is important in the capacity of cells to respond to and cope with changes in their environment.Specifications
P36222 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
This ELISA pair antibody detects human CHI3L1. Other species not determined. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
1116 | |
<10% | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |
6 pg/mL | |
Biotin | |
Kinase & Inhibitors | |
Human | |
Colorimetric Microplate Reader | |
<12% | |
HRP | |
Angiogenesis | |
Human | |
4 hr. 45 min. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.