Learn More
Invitrogen™ CHIA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593057
Description
Immunogen sequence: MVSTPENRQT FITSVIKFLR QYEFDGLDFD WEYPGSRGSP PQDKHLFTVL VQEMREAFEQ EAKQINKPRL MVTAAVAAGI SNIQSGYEIP QLSQYLDYIH VMTYDLHGSW EGYTGENSPL YKYPTDTGSN AYLNVDYVMN YWKDNGAPAE KLIVGFPTYG HNFILSNPSN TGIGAPTSGA GPAGPYAKES GIWAYYEICT FLKNGATQGW DAPQEVPYAY QGNVWVGYDN IKSFDIKAQW LKHNKFGGAM VWAIDLDDFT GTFCNQGKFP LISTLKKALG LQSASCTAPA QPIEPITAAP SGSGNGSGSS SSGGSSGGSG FCAVRANGLY PVANNRNAFW HCVNGVTYQQ NCQAGLVFDT SCDCCNWA; Positive Samples: U-87MG, HepG2, Mouse stomach, Rat lung, Rat stomach; Cellular Location: Cytoplasm.
CHIA degrades chitin and chitotriose. May participate in the defense against nematodes and other pathogens. There are 3 named isoforms produced by alternative splicing. CHIA is induced via a T helper-2 (Th2)-specific, interleukin-13-mediated pathway in epithelial cells and macrophages. CHIA may be an important mediator of IL13-induced responses in Th2-dominated disorders such as asthma. CHIA hydrolysis N-acetyl-beta-D-glucosaminide 1,4-beta-linkages in chitin and chitodextrins.
Specifications
CHIA | |
Polyclonal | |
Unconjugated | |
CHIA | |
2200003E03Rik; acidic chitinase; acidic mammalian chitinase; AMCASE; Chia; Chia1; CHIT2; chitinase, acidic; chitinase, acidic 1; eosinophil chemotactic cytokine; lung-specific protein TSA1902; small acid mammalian chitinase; TSA1902; YNL | |
Rabbit | |
Affinity Chromatography | |
RUO | |
113901, 27159, 81600 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.06 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6RY07, Q91XA9, Q9BZP6 | |
CHIA, Chia1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-368 of human CHIA (NP_0012449321). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.