Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chicago Sky Blue 6B

C.I. 24410

Manufacturer:  Alfa Aesar A1424214

Quantity 25g

 View more versions of this product

Catalog No. AAA1424214


  Check Availability
Add to cart

Specifications

Description

Solubility
Soluble in water (1 mg/ml).

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Description & Specifications

Chemical Identifiers

2610-05-1
992.79
Pontamine Sky Blue 6BX
C34H24N6Na4O1
MFCD00004020

Specifications

2610-05-1
992.79
25g
C34H24N6Na4O1
MFCD00004020
Pontamine Sky Blue 6BX

Safety and Handling

GHS H Statement
H350

GHS P Statement
P201-P281-P202-P308+P313-P405-P501a

Danger

RTECSNumber : QJ6430000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit