Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AU1 tag, Chicken, Polyclonal Antibody, Abnova™

Chicken polyclonal antibody raised against AU1 tag.

Manufacturer:  Abnova Corporation PAB29201

Catalog No. 89-138-540

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Sequence: DTYRYI
Specifications

Specifications

AU1 tag
Unconjugated
ELISA (Capture: 1:200-1:500) Immunohistochemistry (1:250-1:1000) Western Blot (1:1000-1:10000) The optimal working dilution should be determined by the end user.
Liquid
Asynthetic peptide corresponding to AU1 tag and coupled to KLH.
Antigen affinity purification
RUO
Primary
Human
ELISA, Immunohistochemistry, Western Blot
Chicken polyclonal antibody raised against AU1 tag.
In PBS, pH 7.2 (0.09% sodium azide)
Chicken
IgY
100 ug
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only