Learn More
HIS peptide, Chicken, HRP, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against recombinant HIS peptide.
Manufacturer:  Abnova Corporation PAB29139
Catalog No. 89-138-507
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
HIS peptide
HRP
ELISA (1:5000) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide (conjugated with KLH) corresponding to 6X-Histidine tag.
Affinity purification
RUO
Primary
Not Provided
ELISA, Western Blot
Chicken polyclonal antibody raised against recombinant HIS peptide.
In 25mM HEPES, pH 7.0 (0.05M NaCl, 0.1% BSA, 50% glycerol, 0.005% thimerosal).
Chicken
IgY
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal