Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chicken anti-Human IgM Secondary Antibody, Invitrogen™
Chicken Polyclonal Secondary Antibody
Supplier: Thermo Scientific SA172050
Description
SA1-72050 detects Human IgM in human samples. SA1-72050 has been successfully used in ELISA and western blot procedures. The SA1-72050 immunogen is from purified pooled normal human IgM.
Human IgM Polyclonal antibody specifically detects Human IgM in Human samples. It is validated for ELISA, Western Blot.Specifications
Human IgM | |
Polyclonal | |
Unconjugated | |
Chicken | |
Affinity chromatography | |
RUO | |
Human | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.075% sodium azide; pH 7.2 | |
Purified pooled normal human IgM. | |
500 μg | |
Secondary | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgY |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction