Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgA, Chicken, Polyclonal Antibody, Abnova™

Chicken polyclonal antibody raised against native Human IgA.

Manufacturer:  Abnova Corporation PAB29180

Catalog No. 89-138-533

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

IgA
Unconjugated
ELISA (Capture: 1:100-1:500, Detection: 1:5000-1:30000) Western Blot (1:2000-1:10000) The optimal working dilution should be determined by the end user.
Liquid
Native purified IgA from human serum.
Antigen affinity purification
RUO
Primary
Human
ELISA, Western Blot
Chicken polyclonal antibody raised against native Human IgA.
In PBS, pH 7.2 (0.09% sodium azide)
Chicken
IgY
500 ug
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only