Learn More
Chicken IgG, F(ab), Biotin, Isotype control, Novus Biologicals™
Chicken Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197164
Catalog No. NBP197164
Add to cart
Description
Description
IgG F(ab) antibody specifically detects IgG F(ab) in Chicken samples. It is validated for SDS-Page.
Specifications
Specifications
IgG F(ab)
LYOPH
SDS-Page 1:100 - 1:2000
Conjugated
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Chicken IgG, anti-Chicken IgG F(ab)2 and anti-Chicken Serum. No reaction was observed against anti-Chicken IgG F(c) or anti-Papain.
Chicken
SDS-Page
Biotin
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
Chicken
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.