Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chicken IgG, F(ab), Texas Red, Isotype control, Novus Biologicals™

Chicken Isotype Control

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197165

Catalog No. NBP197165


Add to cart

Description

Description

IgG F(ab) antibody specifically detects IgG F(ab) in Chicken samples. It is validated for SDS-Page.
Specifications

Specifications

IgG F(ab)
LYOPH
SDS-Page 1:100 - 1:2000
Conjugated
IgG
1 mg
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Chicken IgG, anti-Chicken IgG F(ab)2 and anti-Chicken Serum. No reaction was observed against anti-Chicken IgG F(c) or anti-Papain.
Chicken
SDS-Page
Texas Red
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
Chicken
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit