Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chicken IgG, H/L Chains, Agarose Immobilized, Isotype control, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197258

Catalog No. NBP197258


Add to cart

Description

Description

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Chicken samples. It is validated for Immunoprecipitation.
Specifications

Specifications

IgG (H+L)
Agarose
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Goat
IgG
20 mg
Store at 4C in the dark.
Polyclonal
Immunoprecipitation
Immunoprecipitation
Conjugated
Chicken IgG whole molecule
IgG purified
RUO
Secondary
Chicken
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit