Learn More
Kingfisher Biotech Inc Chicken LIF Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1395C005
Encompass
The Chicken Leukemia Inhibitory Factor yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken Leukemia Inhibitory Factor applications are for cell culture, ELISA standard, and Western Blot Control. Chicken Leukemia Inhibitory Factor yeast-derived recombinant protein can be purchased in multiple sizes. Chicken Leukemia Inhibitory Factor Specifications: (Molecular Weight: 21.0 kDa) (Amino Acid Sequence: RALLGTSSAC PTNGLCRANV LEQTRRQVAL LNATAQDLFS LYLKCQGEPF SSESDRLCSP SGIFFPPFHV NRTTERKEVM VAMYKLFAFL NASLGNITRD QEELNPMAKE LLNRLHNTTK TTRGLISNLT CLLCKHYNIF QVDVSYGESS KDKSAFKKKQ QGCQVLRKYV QVIAQAARVL LPHLSPA (187)) (Gene ID: 427718). For research use only. Made in the USA
Catalog No. 50-253-3313
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.