Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Chicken Lysozyme ELISA Kit (Colorimetric)

High sensitivity and specificity for detection of chicken lysozyme.

$602.50

Specifications

Product Type ELISA Kit (Colorimetric)
Assay Range 15.625 to 4000ng/mL
Species Chicken
Sensitivity 14.876ng/mL
Sample Type serum, plasma and egg white
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600881KT View Documents Novus Biologicals™
NBP2600881KIT
1 Kit Each for $602.50
Add to cart
 
Description

Description

The Chicken Lysozyme ELISA Kit (Colorimetric) measures chicken Lysozyme in serum, plasma and egg white.
Specifications

Specifications

ELISA Kit (Colorimetric)
Chicken
serum, plasma and egg white
RUO
For detection of chicken lysozyme
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
Adhesive Strip (For 96 wells), Assay plate, Biotin-conjugate (100 x concentrate), Biotin-conjugate Diluent, HRP-avidin Diluent, HRP-avidin (100 x concentrate), TMB Substrate, Wash Buffer (25 x concentrate), Stop Solution
15.625 to 4000ng/mL
14.876ng/mL
Cardiovascular Biology
50 to 100μL
EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis
Lysozyme
ELISA

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit